VWR® Cryogenic Fiberboard Freezer Box
Catalog # 89214-758
Supplier: VWR International
Small Business Enterprise
Specifications
- Description:2" Cryogenic mini box, with drain slots, with grids
- Dimensions:7.4L×7.4W×5.2H cm (2⁹/₁₀×2⁹/₁₀×2¹/₂₀")
- Cat. no.:89214-758
- Cell Dimensions (W×L):1.3 cm (0.53")
Specifications
About this item
Fiberboard boxes can withstand LN₂ storage and do not deteriorate wtihin the LN₂, breakdown can occur if the boxes are removed from the LN₂ and left to condensate in ambient temperatures.
- Multiple heights of boxes to accommodate a variety of heights of vials/tubes (purchase divider wtih empty box)
- Printed alignment arrows (fiberboard boxes with grids only)
- Drain slots for LN₂ drainage (fiberboard boxes with with grids only)
Customers Who Bought This Also Bought