VWR® Cryogenic Fiberboard Freezer Box
# de catalogue 89214-754
Fournisseur: VWR International
Small Business Enterprise
Spécifications
- Dimensions:13.2L×13.2W×5.1H cm (5¹/₅×5¹/₅×2")
- Description:2" Cryogenic box, with drain slots with printed 100 cell grid on lid, no divider
- Cat. no.:89214-754
- Cell Dimensions (W×L):1.3 cm (0.5")
Spécifications
A propos de cet article
Fiberboard boxes can withstand LN₂ storage and do not deteriorate wtihin the LN₂, breakdown can occur if the boxes are removed from the LN₂ and left to condensate in ambient temperatures.
- Multiple heights of boxes to accommodate a variety of heights of vials/tubes (purchase divider wtih empty box)
- Printed alignment arrows (fiberboard boxes with grids only)
- Drain slots for LN₂ drainage (fiberboard boxes with with grids only)
Customers Who Bought This Also Bought