ContactUsLinkComponent
2 inch Mechanical Cryogenic Freezer Box, No Divider, Ea. 5 1/4 inch X 5 1/4 inch X 2 inch
VWR® Cryogenic Fiberboard Freezer Box
# de catalogue 89214-750
Fournisseur:  VWR International
Small Business Enterprise
2 inch Mechanical Cryogenic Freezer Box, No Divider, Ea. 5 1/4 inch X 5 1/4 inch X 2 inch
VWR® Cryogenic Fiberboard Freezer Box
# de catalogue 89214-750
Fournisseur:  VWR International
Small Business Enterprise
Spécifications
  • Dimensions:
    13.2L×13.2W×5.1H cm (5¹/₅×5¹/₅×2")
  • Description:
    2" Cryogenic box, with drain slots, no grids, no divider
  • Cat. no.:
    89214-750
  • Cell Dimensions (W×L):
Spécifications
A propos de cet article
Fiberboard boxes can withstand LN₂ storage and do not deteriorate wtihin the LN₂, breakdown can occur if the boxes are removed from the LN₂ and left to condensate in ambient temperatures.
  • Multiple heights of boxes to accommodate a variety of heights of vials/tubes (purchase divider wtih empty box)
  • Printed alignment arrows (fiberboard boxes with grids only)
  • Drain slots for LN₂ drainage (fiberboard boxes with with grids only)
Customers Who Bought This Also Bought