Black Phenolic Dropper Assembly, Qorpak®
# de catalogue 89210-112
Fournisseur: Qorpak
Small Business Enterprise
Spécifications
- Cap Size:20 mm
- Size:20 mm
- Cat. no.:89210-112
- Supplier No.:CAP-05742
Spécifications
A propos de cet article
Dropper assemblies feature a glass tube with black polypropylene closure and rubber squeeze bulb
Tip opening has a 3.0 mm outside diameter.
Customers Who Bought This Also Bought