ContactUsLinkComponent
ENDURA CHEM COMP DUO 12 RXN
Endura™ Competent Cells, Biosearch Technologies
# de catalogue 71003-028
Fournisseur:  Lucigen
ENDURA CHEM COMP DUO 12 RXN
Endura™ Competent Cells, Biosearch Technologies
# de catalogue 71003-028
Fournisseur:  Lucigen
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Endura Chemically Competent Cells (≥1 × 10^8 cfu/µg DNA)
  • Size:
    12 reactions
  • Genotypes:
    recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ– leu mtl-1 F– mcrB mrr hsdS20(rB–, mB–)
  • Cat. no.:
    71003-028
  • Packaging:
    DUOs
  • Supplier No.:
    60240-1
Spécifications
A propos de cet article
With Lucigen’s Endura Competent cells, you can clone unstable sequences with exceptionally reliable results
Lucigen’s Endura Competent Cells are optimal for conserving clones that contain unstable sequences and maintaining unstable DNA. Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Lucigen offers your choice of two packaging formats and noteably high efficiencies: over 1 × 10^8 cfu/µg (chem) or 1 × 10^10 cfu/µg (electro).
Lucigen’s Endura Competent Cells provide superior value and efficiency. Switch to Endura cells today and experience a higher level of efficiency and a savings in your cloning budget.
Available as chemically competent or electrocompetent cells, Endura Competent Cells are an excellent value and are direct replacements for Invitrogen Stbl3™ and Agilent SURE strains.
Ordering information: Endura Competent Cells include Control DNA and Recovery Medium, and are packaged as SOLOs (1 transformation per tube) or DUOs (2 transformations per tube) as indicated. Endura Electrocompetent cells are provided in DUOs format only. Recovery Medium is also available separately. The specified transformation efficiencies are with pUC DNA, unless indicated otherwise.
Customers Who Bought This Also Bought