Spécifications
- Description:Endura Chemically Competent Cells (≥1 × 10^8 cfu/µg DNA)
- Size:12 reactions
- Genotypes:recA13 supE44 ara-14 galK2 lacY1 proA2 rpsL20(StrR) xyl-5 λ– leu mtl-1 F– mcrB mrr hsdS20(rB–, mB–)
- Cat. no.:71003-028
- Packaging:DUOs
- Supplier No.:60240-1
Spécifications
A propos de cet article
With Lucigen’s Endura Competent cells, you can clone unstable sequences with exceptionally reliable results
Lucigen’s Endura Competent Cells are optimal for conserving clones that contain unstable sequences and maintaining unstable DNA. Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Lucigen offers your choice of two packaging formats and noteably high efficiencies: over 1 × 10^8 cfu/µg (chem) or 1 × 10^10 cfu/µg (electro).
Lucigen’s Endura Competent Cells provide superior value and efficiency. Switch to Endura cells today and experience a higher level of efficiency and a savings in your cloning budget.
Available as chemically competent or electrocompetent cells, Endura Competent Cells are an excellent value and are direct replacements for Invitrogen Stbl3™ and Agilent SURE strains.