20 L Jacketed Filter Reactor System, Electric Motor, Chemgalss
Catalog # 89209-354
Supplier: Chemglass
Small Business Enterprise
Specifications
- Capacity:20 L
- Description:Jacketed Cylindrical Process Filter Reactor System Complete
- Cat. no.:89209-354
- Motor Type:Electric motor
- Supplier No.:CG-1959-F620
Specifications
About this item
Jacketed filter glass reactor features single or multi-step.
- Complete reactor system on open air, autocenter stand
- 150 °C maximum temperature limit for filter base
- Filter base eliminates material transfers and significantly reduces losses
Can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.
Customers Who Bought This Also Bought