PIG® Poly Drip Decks, New Pig
# de catalogue 89208-792
Fournisseur: 2 D 2 S.A. DE C.V. MX
Spécifications
- Description:Holds Four 55-gal. steel or poly drums
- Cat. no.:89208-792
- Load Capacity:2721.6 kg (6000 lbs.)
- Sump Capacity:166.5 L (44 gal.)
- Supplier No.:PAK566
Spécifications
A propos de cet article
Low-profile 5.75"H design is easy to load or unload; pumps and funnels are easy to access
- Low-profile, 5.75"H design is easy to load or unload; pumps and funnels easy to access
- Molded-in sump catches leaks, drips and spills, keeps storage areas clean and safe
- Low-density polyethylene (LDPE) resists UV rays, rust, corrosion and most chemicals
Textured grating adds traction and lifts out for easy sump access. Translucent white sidewalls allow easy visual inspection.
Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe. Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals.
Customers Who Bought This Also Bought