Corning® Clear Bottom, Black and White, Polystyrene Microplates, 384 Well, Corning
# de catalogue 89207-930
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Sterility:Non sterile
- Well Volume:112 µl
- Color:Black with Clear Bottom
- Description:Corning® 384-well, low flange, polystyrene, without lid
- Environmentally Preferable:Low Manufacturing Impact
- No. of wells:384
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Packaging:25/Bag
- Well Shape:Flat-Bottom
- Surface Treatment:Untreated
- Supplier No.:3762
Spécifications
A propos de cet article
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Customers Who Bought This Also Bought