ContactUsLinkComponent
Stylish 2 Lens Spectacle
VWR® Protective Eyewear with Adjustable Temples
# de catalogue 89187-984
Fournisseur:  VWR International
CSA Certified
Product available on GSA Advantage®
Stylish 2 Lens Spectacle
VWR® Protective Eyewear with Adjustable Temples
# de catalogue 89187-984
Fournisseur:  VWR International
CSA Certified
Product available on GSA Advantage®
Spécifications
  • Lens Coating:
    Anti-fog, scratch-resistant
  • Lens Tint:
    Clear
  • Frame Color:
    Black
Spécifications
A propos de cet article
Ultra-lightweight nylon frame with soft PVC nose pads.
  • Unique ratcheting/adjustable temples lock into place for a custom fit
  • Ultra-lightweight nylon frames and soft PVC nose pads to prevent slipping
  • 8-base spherical lenses provide excellent side protection
  • Anti-fog coating extends lens life
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection. The impact resistant polycarbonate lenses provide 99.9% UV protection and the scratch resistant coating.
Certifications: ANSI Z87.1
Customers Who Bought This Also Bought