VWR® Protective Eyewear with Adjustable Temples
# de catalogue 89187-984
Fournisseur: VWR International
CSA Certified
Product available on GSA Advantage®
Spécifications
- Lens Coating:Anti-fog, scratch-resistant
- Lens Tint:Clear
- Frame Color:Black
Spécifications
A propos de cet article
Ultra-lightweight nylon frame with soft PVC nose pads.
- Unique ratcheting/adjustable temples lock into place for a custom fit
- Ultra-lightweight nylon frames and soft PVC nose pads to prevent slipping
- 8-base spherical lenses provide excellent side protection
- Anti-fog coating extends lens life
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection. The impact resistant polycarbonate lenses provide 99.9% UV protection and the scratch resistant coating.
Certifications: ANSI Z87.1
Customers Who Bought This Also Bought