ART® Essentials Kit, MBP
# de catalogue 89204-830
Fournisseur: Thermo Scientific
Spécifications
- Volume:10 µl, 200 µl, 1000 µl
- Tip Color:Clear
- Packed:96 Tips/tray, 12inserts/Pack
- Filter:+
- Tip Style:ART® 10 REACH, ART 200®, ART 1000XL, w/ART® barrier, Reload Inserts
- Supplier No.:2931-RI
Spécifications
A propos de cet article
The ART® Essentials Kit includes the three most popular tip volumes as one complete kit.
Choose from the ART® reload System's hinged racks or reload inserts and protect your pipette from aerosol and liquid contaminiantion with the original aerosol resistent barrier tip.
Customers Who Bought This Also Bought