Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
# de catalogue 16171-819
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Material:Blue Aluminum
- Cat. no.:16171-819
- For O.D.:13 mm
- Supplier No.:224182-05
Spécifications
A propos de cet article
One-piece center disc tears out, exposing top of seal to syringe
Shelf packs of 100.
Customers Who Bought This Also Bought