VWR® Lens Cleaning Wipes and Lens Cleaning Station
Catalog # 89203-548
Supplier: VWR International
Specifications
- Description:VWR® Disposable lens cleaning station
- Cat. no.:89203-548
Specifications
About this item
VWR® lens cleaning wipes and lens cleaning station provide a clean, safe, and effective source of personal protection that can be conveniently positioned virtually anywhere in the workplace.
- Cleans without scratching, streaking, or residue
- Individually foil wrapped, premoistened
- Disposable station contains cleaning solution and dry cleaning tissues
- Solution coats lenses with anti-fog, anti-static formula
Dispenser box is convenient and portable for use anywhere, anytime.
The solution cleans and coats the lens, and will not leave any excess liquid on the lens surface. Disposable stations can be positioned close to workers and easily moved.
Not for use on contact lenses or specially coated optical lenses, such as anti-glare coatings.
Ordering information: The 17.8×12.7 cm (7×5") lens cleaning wipes are individually foil-packed and pre-saturated with lens cleaning solution. Packaged 120 wipes per box. Lens cleaning station contains one 3.5 oz. bottle of lens cleaning solution and one pack of 15.2×12.7 cm (6×5") heavy-duty lens cleaning tissues.
Delivery: The lens cleaning station includes one bottle of special anti-fog, anti-static solution and one pack of heavy-duty lens cleaning tissues.
Frequently Bought Together