Order Entry
ContactUsLinkComponent
VWR SWAB 6IN COTTON TIP PK1000
VWR® Critical Swab® Swab, Cotton Head
Catalog # 89031-270
Supplier:  VWR International
Product available on GSA Advantage®
VWR SWAB 6IN COTTON TIP PK1000
VWR® Critical Swab® Swab, Cotton Head
Catalog # 89031-270
Supplier:  VWR International
Product available on GSA Advantage®

Specifications

  • Cat. no.:
    89031-270
  • Handle Length:
    152.0 mm (6")
  • Handle Width:
    2.5 mm (0.1")
  • Head Length:
    15.9 mm (0.624")
  • Head Width:
    4.8 mm (0.188")

Specifications

About this item

Disposable swab with absorbent cotton tip and wood shaft.

  • Autoclavable
  • Disposable
Customers Who Bought This Also Bought