ContactUsLinkComponent
Spreader T Curved 150Mm Cs3500
Corning® Gosselin™ Spreaders
# de catalogue 89200-968
Fournisseur:  Corning
Spreader T Curved 150Mm Cs3500
Corning® Gosselin™ Spreaders
# de catalogue 89200-968
Fournisseur:  Corning
Spécifications
  • Length:
    150 mm
  • Sterility:
    Sterile
  • Color:
    Green
  • Description:
    T-shape
  • Cat. no.:
    89200-968
  • Packaging:
    25/Bag, 28 Bags/Box
  • Supplier No.:
    ETAT-04
Spécifications
A propos de cet article
These spreaders are made of PS.
  • Curved end or "T" end
  • Sterile
They are packed in a tamper-evident resealable bag.
Customers Who Bought This Also Bought