ROTOR SMALL
# de catalogue WT41-1499
Fournisseur: Welch Vacuum
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$146.85 to $576.51
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Fournisseur: MilliporeSigma
$695.79
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
$73.48 to $4,408.00
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
$775.79 to $2,413.44
Nalgene® Polyethylene Dewar Flask, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$994.67