INNER RING
# de catalogue WT41-1042
Fournisseur: Welch Vacuum
Customers Who Bought This Also Bought
Sulfuric acid 2.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
119,71 $ to 1 363,18 $
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
277,09 $ to 740,95 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Fournisseur: VWR International
131,08 $ to 405,43 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
377,44 $ to 992,90 $