Standard Taper Glass Stopper with KEM-KLAMP® Lug, Kimble Chase
# de catalogue KT851800-2440
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- ST Joint No.:24/40
- Cat. no.:KT851800-2440
- Supplier No.:851800-2440
Spécifications
A propos de cet article
Designed for use with 675500 series KEM-KLAMPS®
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Pennyhead top has a lug attachment to engage the spring portion of the KEM-KLAMP®.
Customers Who Bought This Also Bought