ContactUsLinkComponent
Stoppers Glass Stopper Solid [standard taper] Pennyhead with Lug KEM-KLAMP
Standard Taper Glass Stopper with KEM-KLAMP® Lug, Kimble Chase
# de catalogue KT851800-2440
Stoppers Glass Stopper Solid [standard taper] Pennyhead with Lug KEM-KLAMP
Standard Taper Glass Stopper with KEM-KLAMP® Lug, Kimble Chase
# de catalogue KT851800-2440
Spécifications
  • ST Joint No.:
    24/40
  • Cat. no.:
    KT851800-2440
  • Supplier No.:
    851800-2440
Spécifications
A propos de cet article
Designed for use with 675500 series KEM-KLAMPS®
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Pennyhead top has a lug attachment to engage the spring portion of the KEM-KLAMP®.
Customers Who Bought This Also Bought