ContactUsLinkComponent
Stopper, Standard Taper Pennyhead, Medium Length
KONTES® Pennyhead [ST] Stoppers, Medium Length, Kimble Chase
# de catalogue KT850500-1018
Stopper, Standard Taper Pennyhead, Medium Length
KONTES® Pennyhead [ST] Stoppers, Medium Length, Kimble Chase
# de catalogue KT850500-1018
Spécifications
  • Length:
    18 mm (0.71")
  • ST Joint No.:
    10/18
  • Cat. no.:
    KT850500-1018
  • Supplier No.:
    850500-1018
Spécifications
A propos de cet article
Glass pennyhead stoppers
Sizes [ST]10/18 through [ST]19/22 are solid. Sizes [ST]24/25 and larger are hollow.
Customers Who Bought This Also Bought