Medium Length Hex Head Hollow Glass Stoppers, Kimble Chase
# de catalogue KT850400-2926
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- ST Joint No.:29/26
- Cat. no.:KT850400-2926
- Supplier No.:850400-2926
Spécifications
A propos de cet article
Stoppers are used with laboratory glassware such as flasks, mixing cylinders, and separatory funnels
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Hollow, hexagonal head medium length Standard Taper stopper can be easily gripped, and may be set on the flattened top to avoid contamination of the ground surface.
Customers Who Bought This Also Bought