KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
# de catalogue KT617000-0624
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:500 ml
- ST Joint No.:24/40
- Supplier No.:617000-0624
Spécifications
A propos de cet article
Single neck flask with a standard taper outer joint.
- White ceramic enamel marking spot
- Manufactured from 33 expansion, low extractable borosilicate glass
Tops are reinforced and tooled with a rounded finish, containing more glass to give them maximum mechanical strength. Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought