ContactUsLinkComponent
Chromatography VALVES
One-Way Stopcock Valves, Female to Male, Kimble Chase
# de catalogue KT420163-4500
Chromatography VALVES
One-Way Stopcock Valves, Female to Male, Kimble Chase
# de catalogue KT420163-4500
Spécifications
  • Color:
    Blue
  • Description:
    One-way stopcock valve, nylon body, HDPE plug, female luer to male luer
  • Cat. no.:
    KT420163-4500
  • Supplier No.:
    420163-4500
Spécifications
A propos de cet article
One-way stopcocks with a female to male connection and HDPE plug.
Customers Who Bought This Also Bought