ContactUsLinkComponent
CUSTODY SEALS,1" X 7
Custody Seals, Thermo Scientific
# de catalogue IR500
Fournisseur:  Thermo Scientific
CUSTODY SEALS,1" X 7
Custody Seals, Thermo Scientific
# de catalogue IR500
Fournisseur:  Thermo Scientific
Spécifications
  • Dimensions:
    2.5L x 17.8W cm (1 x 7")
  • Description:
    Custody Seals - Adhesive Paper Labels
  • Cat. no.:
    IR500
  • Supplier No.:
    500
Spécifications
A propos de cet article
Custody Seals provide complete chain-of-custody and tamper evidence for samples from the point of collection to lab receipt.
Custody seal adhesive stickers securely seal sample bottles and cannot be peeled off. The only way to open the sealed bottle is to break the paper label. Each seal includes a writing area for custody signature and date for tamper-evident secutity. Apply at the point of sample collection, and ship samples to their destination with confidence.
Customers Who Bought This Also Bought