Potassium iodide ≥99.0%, crystals, GR ACS, Supelco®
# de catalogue CAPX1505-1
Fournisseur: MilliporeSigma
Numéro CAS: 7681-11-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:500 g
- Cat. no.:CAPX1505-1
- Supplier No.:PX1505-1
Spécifications
Résultats des tests de spécification
- Assay:99.0% min
- Barium (Ba):0.002% max
- Calcium (Ca):0.002% max
- Chloride and bromide (as Cl):0.01% max
- Color:White
- Form:Crystals or fine granules
- Heavy metals (as Pb):5ppm max
- Insoluble matter:0.005% max
- Iodate (IO3):3ppm max
- Iron (Fe):3ppm max
- Loss on drying (150°C):0.2% max
- Magnesium (Mg):0.001% max
- pH of a 5% solution at 25°C:6.0-9.2
- Phosphate (PO4):0.001% max
- Sodium (Na):0.005% max
- Sulfate (SO4):0.005% max
- Supelco®, Potassium iodide ≥99.0%, crystals, GR ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Dipotassium diiodide
Point d'ébullition:1330 °C (1013 hPa)
Point de Fusion:686 °C
Densité:3,13 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Point d'ébullition:
:Pt ébul.: 1330 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: 686 °C
:Point eclaire:
:Flash Pt:
:M.W. 166 g/mol
Index Merck:13,07727
EINECS:231-659-4
:686
:1330
:GHS08
Température d'entreposage:Température ambiante
:12,07809
Customers Who Bought This Also Bought