Petroleum spirit, 35…60 °C, OmniSolv®, Supelco®
# de catalogue CAPX0424-1
Fournisseur: MilliporeSigma
Numéro CAS: 64742-49-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAPX0424-1
- Supplier No.:PX0424-1
Spécifications
Résultats des tests de spécification
- Boiling range:35-60°C
- Capillary ECD responsive substances (as PCNB):2ng/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:1ppm max
- Titrable acid:0.2µeq/g max
- Water (H2O):0.01% max
- Petroleum spirit, 35…60 °C, OmniSolv®, Supelco®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Gasoline,Petroleum ether
:Length:
:Width:
:Height:
:Ø
:Thickness:
:35…60 °C
:Boiling Pt:
:Boiling Pt: 100 to 120 °C (1013 hPa)
:Melting Pt:
:Flash Pt:
:Flash Pt: -10 °C
:Density: 0.7 g/cm³ (20 °C)
:0.7
:-10
Customers Who Bought This Also Bought