Methanol ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
# de catalogue CAMX0486-1
Fournisseur: MilliporeSigma
Numéro CAS: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAMX0486-1
- Supplier No.:MX0486-1
Spécifications
Résultats des tests de spécification
- Assay (GC):99.9% min
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence as quinine at 254 nm:1ppb max
- Fluorescence as quinine at 365 nm:1ppb max
- Form:Clear liquid
- Gradient at 235 nm:1mAU max
- Gradient at 254 nm:1mAU max
- Identity (IR-spectrum):Conforms
- Metals, suitable for LC/MS:To pass test
- MS purity, ESI(+), largest response as reserpine:50ppb max
- MS purity, ESI(-), largest response, p-nitrophenol:50ppb max
- Residue after evaporation:1ppm max
- Titrable acid:0.3µeq/g max
- Titrable base:0.2µeq/g max
- UV Abs. at 210 nm:0.3AU max
- UV Abs. at 220 nm:0.1AU max
- UV Abs. at 230 nm:0.04AU max
- UV Abs. at 240 nm:0.03AU max
- UV Abs. at 250 nm:0.01AU max
- UV Abs. at 270 nm:0.01AU max
- UV Abs. at 400-280 nm:0.005AU max
- UV Cut-off:205nm max
- Water (H2O):0.05% max
- Supelco®, Methanol ≥99.9%, OmniSolv® LC-MS for LC-MS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxy methane,Carbinol,Wood alcohol
Point d'ébullition:64,6 °C (1013 hPa)
Point de Fusion:-98 °C
Densité:0,7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
Point d'éclair:11 °C (closed cup)
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 64,6 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -98 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 11 °C (coupe fermée)
:M.W. 32,04 g/mol
Index Merck:13,05984
Point d'ébullition:64.6 °C (1013 hPa)
Point de Fusion:–98 °C
Température d'entreposage:Ambient
EINECS:200-659-6
:-98
:64,6
Densité:0.7918 g/cm³ (20 °C)
ONU:1230
:01-2119433307-44
:11
:GHS02
:GHS06
:GHS08
Température d'entreposage:Température ambiante
:99,9
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
:3
:II
Customers Who Bought This Also Bought