Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
# de catalogue CAJT9012-3
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 75-05-8
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Supplier No.:9012-03
Spécifications
Résultats des tests de spécification
- For Laboratory,Research,or Manufacturing Use:
- Appearance:Passes Test
- Assay (GC):≥99.9 %
- Color (APHA):≤10
- Water (H₂O):≤0.02 %
- Gradient Elution Test (a.u.) - 254 nm:≤0.005
- Residue after Evaporation:≤3 ppm
- Titrable Acid (µeq/g):≤8
- Titrable Base (µeq/g):≤0.6
- Ultraviolet Absorbance (1.00 cm) - 200 nm:≤0.1
- Ultraviolet Absorbance (1.00 cm) - 210 nm:≤0.05
- Ultraviolet Absorbance (1.00 cm) - 220 nm:≤0.03
- Ultraviolet Absorbance (1.00 cm) - 254 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 280 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - 400 nm:≤0.01
- Ultraviolet Absorbance (1.00 cm) - UV Cut-off, nm:≤190
- Filtered through a 0.2 micron filter.:
- Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
Point d'ébullition:81.6 °C (1013 hPa)
Point de Fusion:–45.7 °C
Densité:0.782 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Management of Change (MOC) category = N
Point d'éclair:2 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 81.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –45.7 °C
:Flash Pt:
:Flash Pt: 2 °C (closed cup)
:M.W. 41.05 g/mol
Index Merck:13,00071
:Acetonitrile
Température d'entreposage:Ambient
EINECS:200-835-2
:-45.7
ONU:1648
:01-2119471307-38
:3
:II
:2
:GHS02
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought