Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
# de catalogue CAHX0603-4
Fournisseur: MilliporeSigma
Numéro CAS: 7647-01-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, pre-packed
- Size:500 mL
- Cat. no.:CAHX0603-4
- Supplier No.:HX0603-4
Spécifications
Résultats des tests de spécification
- Assay:36.5-38.0%
- Aluminum (Al):0.3ppm max
- Ammonium (NH4):3ppm max
- Appearance:To pass test
- Arsenic (As):0.01ppm max
- Bromide (Br):0.005% max
- Calcium (Ca):0.3ppm max
- Chromium (Cr):0.2ppm max
- Color (APHA):10 max
- Copper (Cu):0.1ppm max
- Extractable organic substances (about 5 ppm):To pass test
- Free chlorine (Cl):1ppm max
- Gold (Au):0.3ppm max
- Heavy metals (as Pb):1ppm max
- Iron (Fe):0.2ppm max
- Lead (Pb):0.1ppm max
- Magnesium (Mg):0.3ppm max
- Manganese (Mn):0.3ppm max
- Mercury (Hg):0.005ppm max
- Nickel (Ni):0.1ppm max
- Potassium (K):0.3ppm max
- Residue after ignition:5ppm max
- Sodium (Na):0.3ppm max
- Sulfate (SO4):1ppm max
- Sulfite (SO3):1ppm max
- Tin (Sn):0.3ppm max
- Titanium (Ti):0.3ppm max
- Zinc (Zn):0.3ppm max
- Supelco®, Hydrochloric acid 36.5 - 38.0%, GR ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydrochloric acid solution
Point d'ébullition:110 °C (1013 hPa)
Point de Fusion:–30 °C
Densité:1,18 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
:Point d'ébullition:
:Boiling Pt:
:Point de fusion:
:Pt Fusion: -30 °C
:Point eclaire:
:Flash Pt:
:M.W. 36,46 g/mol
:36.46
Température d'entreposage:Température ambiante
EINECS:231-595-7
ONU:1789
:8
:II
:GHS05
:GHS07
:MQ200
Customers Who Bought This Also Bought