Benzene ≥99.7%, OmniSolv®, Supelco®
# de catalogue CABX0212-1
Fournisseur: MilliporeSigma
Numéro CAS: 71-43-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CABX0212-1
- Supplier No.:BX0212-1
Spécifications
Résultats des tests de spécification
- Assay (GC):99.7% min
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:2ppm max
- UV Abs. at 280 nm:1.00AU max
- UV Abs. at 290 nm:0.15AU max
- UV Abs. at 300 nm:0.05AU max
- UV Abs. at 330 nm:0.01AU max
- UV Abs. at 350 nm:0.005AU max
- UV Cut-off:280nm max
- Water (H2O):0.03% max
Résultats des tests de spécification
A propos de cet article
Synonymes: Cyclohexatriene
Point d'ébullition:80.1 °C (1013 hPa)
Point de Fusion:5.5 °C
Densité:0.878 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
Point d'éclair:–11 °C
:Boiling Pt:
:Boiling Pt: 80.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 5.5 °C
:Flash Pt:
:Flash Pt: -11 °C
:M.W. N/A N/A
Index Merck:13,01066
Température d'entreposage:Ambient
EINECS:200-753-7
ONU:1114
:01-2119447106-44
:3
:II
:-11
:GHS02
:GHS08
Customers Who Bought This Also Bought