N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
# de catalogue CABJ071-4
Fournisseur: Burdick & Jackson
Numéro CAS: 127-19-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CABJ071-4
- Supplier No.:071-4
Spécifications
Résultats des tests de spécification
- GC Analysis:99.5 % min.
- GC Impurities Eluting Before Solvent:0.5 mg/L max.
- UV Absorbance @ 275 nm:0.3 AU max.
- UV Absorbance @ 300 nm:0.08 AU max.
- UV Absorbance @ 350 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:268 nm max.
- Water by Karl Fischer Titration:0.03 % max.
- Burdick & Jackson™, N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry:
Résultats des tests de spécification
A propos de cet article
Synonymes: Acetic acid dimethylamide
Point d'ébullition:166 °C
Point de Fusion:-20 °C
Densité:0.937 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Point d'éclair:70 °C
:Boiling Pt:
:Boiling Pt: 166.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -20 °C
:Flash Pt:
:Flash Pt: 70 °C
:M.W. 87.12 g/mol
Index Merck:13,03254
:N,N-Dimethylacetamide
Température d'entreposage:Ambient
EINECS:204-826-4
:-20
:166.1
ONU:2810
:70
:GHS08
:GHS07
Customers Who Bought This Also Bought