ContactUsLinkComponent
Single Use Luer-Lok* Tip Syringe/PrecisionGlide* Needle Combination
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
# de catalogue CABD309580
Fournisseur:  BD
Single Use Luer-Lok* Tip Syringe/PrecisionGlide* Needle Combination
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
# de catalogue CABD309580
Fournisseur:  BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    BD Luer-Lok Tip
  • Sterility:
    Sterile
  • Cat. no.:
    CABD309580
  • Packaging:
    100/Box, 800/Case
  • Needle Length:
    11/2"
  • Gauge:
    18
  • Supplier No.:
    309580
Spécifications
A propos de cet article
Sterile syringe/needle combinations have Slip tip or BD Luer-Lok™ tip.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Sterile syringe/needle combinations have Slip tip or BD Luer-Lok™ tip.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Thin wall needles have a bigger I.D. without any increase in the O.D. Thin wall needles allow the same flow volume as standard-walled needles one gauge larger.
Needles are made of stainless steel with translucent, color-coded polypropylene hubs.
Caution: Needles are autoclavable after being removed from their easy-peel package. They can be sterilized by ethylene oxide in their easy-peel package.
Customers Who Bought This Also Bought