Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
# de catalogue CAAX0116-1
Fournisseur: MilliporeSigma
Numéro CAS: 67-64-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CAAX0116-1
- Supplier No.:AX0116-1
Spécifications
Résultats des tests de spécification
- Assay (GC, corrected for water content):99.5% min
- Capillary ECD responsive substances (as PCNB):2ng/L max
- Capillary FID responsive substances (as decane):3µg/L max
- Color (APHA):10 max
- Residue after evaporation:1ppm max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):1.0ppb max
- Infrared spectrum:To Pass Test
- Titrable acid:0.3µeq/g max
- UV Abs. at 329 nm:1.00AU max
- UV Abs. at 330 nm:0.80AU max
- UV Abs. at 335 nm:0.30AU max
- UV Abs. at 340 nm:0.06AU max
- UV Abs. at 350 nm:0.005AU max
- UV Abs. at 400 nm:0.005AU max
- UV Cut-off:329nm max
- Water (H2O):0.5% max
- Acetone ≥99.5%(by GC, corrected for water content), OmniSolv®, Supelco®:
Résultats des tests de spécification
A propos de cet article
Synonymes: 2-Propanone,DMK,Propanone
Point d'ébullition:56.2 °C (1013 hPa)
Point de Fusion:–95.4 °C
Densité:0.792 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Point d'éclair:<–20 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: Max. -20 °C (closed cup)
:M.W. 58.08 g/mol
Index Merck:13,00067
EINECS:200-662-2
:-95.4
Température d'entreposage:Ambient
ONU:1090
:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
Customers Who Bought This Also Bought