Acetic acid glacial ≥99.7% for HPLC, Supelco®
Catalog # CAAX0074-6
Supplier: MilliporeSigma
CAS Number: 64-19-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle, pre-packed
- Size:500 ml
- Cat. no.:CAAX0074-6
- Supplier No.:AX0074-6
Specifications
Specification Test Results
- Assay (GC):99.7% min
- Acetic anhydride [(CH3CO)2O]:0.01% max
- Chloride (Cl):0.4ppm max
- Color (APHA):10 max
- Copper (Cu):0.1ppm max
- Dilution test:To pass test
- Form:Clear liquid
- Gradient at 254 nm:0.005AU max
- Gradient at 280 nm:0.003AU max
- Gradient Suitability for LC-MS:Conforms
- Heavy metals (as Pb):0.5ppm max
- Iron (Fe):0.2ppm max
- MS Purity (as Reserpine):Conforms
- Nickel (Ni):0.1ppm max
- Residue after evaporation:5ppm max
- Substances reducing dichromate:To pass test
- Substances reducing permanganate:To pass test
- Sulfate (SO4):0.4ppm max
- Titrable base:0.0004meq/g max
- UV Absorbance (1 cm, 255 nm):1.0AU max
- UV Absorbance (1 cm, 280 nm):0.05AU max
- UV Absorbance (1 cm, 350 nm):0.02AU max
- Water (H2O):0.1% max
- Supelco®, Acetic acid glacial ≥99.7% for HPLC:
Specification Test Results
About this item
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1,05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
Flash Pt:38,5 °C
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 118 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 17 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 38,5 °C
:M.W. 60,05 g/mol
Boiling Pt:118 °C (1013 hPa)
EINECS:200-580-7
:17
:118
UN:2789
REACH:01-2119475328-30
:38,5
:GHS02
:GHS05
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Température ambiante
:99,7
:M-Clarity™ quality level = MQ100
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
:38.5
:MQ100
Storage Temperature:Ambient
Customers Who Bought This Also Bought