Acetic acid glacial ≥99.7%, GR ACS, Supelco®
# de catalogue CAAX0073-6
Fournisseur: MilliporeSigma
Numéro CAS: 64-19-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, pre-packed
- Size:500 ml
- Cat. no.:CAAX0073-6
- Supplier No.:AX0073-6
Spécifications
Résultats des tests de spécification
- Assay (GC):99.7% min
- Acetic anhydride [(CH3CO)2O]:0.01% max
- Aluminum (Al):0.3ppm max
- Arsenic (As):0.05ppm max
- Calcium (Ca):0.3ppm max
- Chloride (Cl):1ppm max
- Chromium (Cr):0.2ppm max
- Color (APHA):10 max
- Copper (Cu):0.1ppm max
- Dilution test:To pass test
- Gold (Au):0.3ppm max
- Heavy metals (as Pb):0.5ppm max
- Iron (Fe):0.2ppm max
- Lead (Pb):0.2ppm max
- Magnesium (Mg):0.3ppm max
- Manganese (Mn):0.2ppm max
- Nickel (Ni):0.1ppm max
- Phosphate (PO4):1.0ppm max
- Potassium (K):0.3ppm max
- Residue after evaporation:0.001% max
- Sodium (Na):0.3ppm max
- Substances reducing dichromate:To pass test
- Substances reducing permanganate:To pass test
- Sulfate (SO4):1ppm max
- Tin (Sn):0.3ppm max
- Titanium (Ti):0.3ppm max
- Titrable base:0.0004meq/g max
- Zinc (Zn):0.3ppm max
- Supelco®, Acetic acid glacial ≥99.7%, GR ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Vinegar acid,Methanecarboxylic acid
Point d'ébullition:118 °C (1013 hPa)
Point de Fusion:17 °C
Densité:1,05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:M-Clarity™ quality level = MQ200
Point d'éclair:38,5 °C
:Point d'ébullition:
:Pt ébul.: 118 °C (1013 hPa)
:Point de fusion:
:Pt Fusion: 17 °C
:Point eclaire:
:Pt Eclair: 38,5 °C
:M.W. 60,05 g/mol
EINECS:200-580-7
:17
:118
Température d'entreposage:Température ambiante
ONU:2789
:01-2119475328-30
:8.3
:II
:38,5
:GHS02
:GHS05
:MQ200
Customers Who Bought This Also Bought