Acetic acid glacial ≥99.7%, GR ACS, Supelco®
Catalog # CAAX0073-59
Supplier: MilliporeSigma
CAS Number: 64-19-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle in plastic box
- Size:2.5 L
- Cat. no.:CAAX0073-59
- Supplier No.:AX0073-59
Specifications
Specification Test Results
- Assay (GC):99.7% min
- Acetic anhydride [(CH3CO)2O]:0.01% max
- Aluminum (Al):0.3ppm max
- Arsenic (As):0.05ppm max
- Calcium (Ca):0.3ppm max
- Chloride (Cl):1ppm max
- Chromium (Cr):0.2ppm max
- Color (APHA):10 max
- Copper (Cu):0.1ppm max
- Dilution test:To pass test
- Gold (Au):0.3ppm max
- Heavy metals (as Pb):0.5ppm max
- Iron (Fe):0.2ppm max
- Lead (Pb):0.2ppm max
- Magnesium (Mg):0.3ppm max
- Manganese (Mn):0.2ppm max
- Nickel (Ni):0.1ppm max
- Phosphate (PO4):1.0ppm max
- Potassium (K):0.3ppm max
- Residue after evaporation:0.001% max
- Sodium (Na):0.3ppm max
- Substances reducing dichromate:To pass test
- Substances reducing permanganate:To pass test
- Sulfate (SO4):1ppm max
- Tin (Sn):0.3ppm max
- Titanium (Ti):0.3ppm max
- Titrable base:0.0004meq/g max
- Zinc (Zn):0.3ppm max
- Supelco®, Acetic acid glacial ≥99.7%, GR ACS:
Specification Test Results
About this item
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1,05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:M-Clarity™ quality level = MQ200
Flash Pt:38,5 °C
:Point d'ébullition:
:Pt ébul.: 118 °C (1013 hPa)
:Point de fusion:
:Melting Pt: 17 °C
:Point eclaire:
:Pt Eclair: 38,5 °C
:M.W. 60,05 g/mol
:GHS02
:GHS05
:8.3
:II
Storage Temperature:Température ambiante
UN:2789
:118
:38,5
:17
EINECS:200-580-7
REACH:01-2119475328-30
:MQ200
Customers Who Bought This Also Bought