di-Ammonium oxalate monohydrate 99.0-101.0% ACS
# de catalogue CAAA36228-22
Fournisseur: Thermo Scientific Chemicals
Numéro CAS: 6009-70-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Bottle
- Size:100 g
- Cat. no.:CAAA36228-22
- Supplier No.:36228-22
Spécifications
Résultats des tests de spécification
- Insoluble matter:0.005%
- Residue after ignition:0.02%
- Cl:0.002%
- SO3:0.002%
- Heavy Metals (as Pb):5 ppm
- Fe:2%
- Color:White
- Form:Granular
- Solubility:Soluble in water, Slightly soluble in alcohol
- di-Ammonium oxalate monohydrate 99.0-101.0% ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Ammonium oxalate monohydrate,Ammonium oxalate-1-hydrate,Oxalic acid diammonium salt monohydrate,Oxalic acid ammonium salt monohydrate,Ammonium ethanedioate monohydrate,Ammonium Oxalate
Point d'ébullition:70 °C (1013 hPa)
Point de Fusion:70 °C (dec.)
Densité:~ 1.5 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Boiling Pt:
:Boiling Pt: 70 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 70 °C (dec.)
:Flash Pt:
:Flash Pt:
:M.W. 142.11 g/mol
Index Merck:13,00541
Température d'entreposage:Ambient
EINECS:214-202-3
:70
ONU:3288
:6.1
:III
:GHS07
:14,537
Customers Who Bought This Also Bought