ContactUsLinkComponent
Bottle Rv Sal Soy Enrichment Broth 500G
Rappaport Vassiliadis Salmonella (RVS) Soy Broth, BD Diagnostics
# de catalogue CA95038-637
Fournisseur:  BD
Numéro CAS:  
Rappaport Vassiliadis Salmonella (RVS) Soy Broth, BD Diagnostics
# de catalogue CA95038-637
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Difco™ Rappaport-Vassiliadis Salmonella (RVS) Soy Broth, Dehydrated
  • Size:
    500 g
  • Cat. no.:
    CA95038-637
  • Supplier No.:
    214943
Spécifications
A propos de cet article
Rappaport-Vassiliadis Salmonella Soy Broth is used for selectively enriching Salmonella in food and environmental samples.
  • Soy peptone is a source of carbon and nitrogen for general growth requirements
  • Malachite green helps inhibit organisms other than Salmonella
  • Low pH helps select for highly resistant Salmonella species
Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) performance specifications, where applicable.
Customers Who Bought This Also Bought