Spécifications
- Description:Difco™ Rappaport-Vassiliadis Salmonella (RVS) Soy Broth, Dehydrated
- Size:500 g
- Cat. no.:CA95038-637
- Supplier No.:214943
Spécifications
A propos de cet article
Rappaport-Vassiliadis Salmonella Soy Broth is used for selectively enriching Salmonella in food and environmental samples.
- Soy peptone is a source of carbon and nitrogen for general growth requirements
- Malachite green helps inhibit organisms other than Salmonella
- Low pH helps select for highly resistant Salmonella species
Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) performance specifications, where applicable.