ContactUsLinkComponent
Diagnostic Reagent and Stain Droppers
BD BBL™ Diagnostic Reagent and Stain Droppers, BD Diagnostics
# de catalogue CA90003-658
Fournisseur:  BD
Numéro CAS:  
Diagnostic Reagent and Stain Droppers
BD BBL™ Diagnostic Reagent and Stain Droppers, BD Diagnostics
# de catalogue CA90003-658
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Oxidase
  • Application:
    Identification of Oxidate-Producing Bacteria
  • Supplier No.:
    261181
Spécifications
A propos de cet article
Reagents are hermetically sealed in an ampule to protect from chemical instability
They are used by holding the dropper upright and squeezing it to crush the ampule inside. The dropper can then be tilted for convenient, drop-by-drop dispensing. Each ampule contains a 1-day supply of premixed reagent and has a shelf life of one to three years without refrigeration. Flat bottoms allow the droppers to stand upright on the benchtop.
Ordering information: Each reagent comes in a box of 50 ampules.
Customers Who Bought This Also Bought