ContactUsLinkComponent
Rosolic Acid
Rosolic Acid, BD
# de catalogue CA90002-602
Fournisseur:  BD
Numéro CAS:  
Rosolic Acid, BD
# de catalogue CA90002-602
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Rosolic Acid
  • Size:
    1 g
  • Cat. no.:
    CA90002-602
  • Supplier No.:
    232281
Spécifications
A propos de cet article
Rosolic Acid is used with m FC Agar and m FC Broth Base in cultivating and enumerating fecal coliforms by the membrane filter technique at elevated temperatures.
  • Practically insoluble in water
  • Freely soluble in alcohol
  • Can be used as a pH indicator
Soluble in strong acids to form yellow solution, or in aqueous alkalis to form carmine red solutions. Because of its behavior in changing color due to the presence of an acidic or alkaline solution, rosolic acid is used as a pH indicator with pH transition range of 5.0-6.8.
Customers Who Bought This Also Bought