Spécifications
- Description:Rosolic Acid
- Size:1 g
- Cat. no.:CA90002-602
- Supplier No.:232281
Spécifications
A propos de cet article
Rosolic Acid is used with m FC Agar and m FC Broth Base in cultivating and enumerating fecal coliforms by the membrane filter technique at elevated temperatures.
- Practically insoluble in water
- Freely soluble in alcohol
- Can be used as a pH indicator
Soluble in strong acids to form yellow solution, or in aqueous alkalis to form carmine red solutions. Because of its behavior in changing color due to the presence of an acidic or alkaline solution, rosolic acid is used as a pH indicator with pH transition range of 5.0-6.8.