ContactUsLinkComponent
Willson A700 Cl Frm, Cl Hc
A700 Series Protective Eyewear, Honeywell Safety
# de catalogue CA89020-064
Willson A700 Cl Frm, Cl Hc
A700 Series Protective Eyewear, Honeywell Safety
# de catalogue CA89020-064
Spécifications
  • Lens Tint:
    Clear
  • Description:
    Lenses with anti-scratch treatment
  • Frame Color:
    Clear
  • Supplier No.:
    A700
Spécifications
A propos de cet article
These economic and sporty glasses provide a stylish fit for the everyday wearer. The lightweight design is equipped with sport temples and soft temple tip pads that offer a secure, wraparound fit.
These economic and sporty glasses provide a stylish fit for the everyday wearer. The lightweight design is equipped with sport temples and soft temple tip pads that offer a secure, wraparound fit. The molded universal nose bridge provides added comfort throughout extended use. The 9-base polycarbonate hardcoated lenses provide a 180° field of distortion-free vision, and offer 99.9% UV protection.
The A700 series is available in a variety of lens styles for both indoor and outdoor work applications. Model 89020-074 features Fog-Ban® treatment to reduce fogging in extreme conditions.All models comply with ANSI Z87+ and CSA Z94.3 standards.
  • Molded universal nose bridge provides added comfort throughout extended use
  • Nine-base polycarbonate hardcoated lenses provide a 180° field of distortion-free vision, and offer 99.9% UV protection
The A700 series is available in a variety of lens styles for both indoor and outdoor work applications. Model 89020-074 features Fog-Ban® treatment to reduce fogging in extreme conditions.
Certifications: All models comply with ANSI Z87+ and CSA Z94.3 standards.
Customers Who Bought This Also Bought