Spécifications
- Size:3.8 L (1 gal.)
- Cat. no.:CA80077-359
- pH:12
- Supplier No.:2101
- Packing Type:Bottle
Spécifications
A propos de cet article
Non ionic, low foaming phosphate free liquid cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. Passes inhibitory residue test.
- Concentrated, biodegradable and readily disposable
- Low-foaming high-wetting detergent for maximum performance
- Ideal for CIP cleaning and floor standing lab washing machines
- Dilute 1:200
For removeing soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals such as stainless steel. Solujet is inhibited for Aluminum. Clean-in place, laboratory warewashers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, and manual cleaning if proper skin and eye protection is used.
Note: Corrosion testing is advisable.
They can be used to clean pharmaceutical process equipment, water testing labware, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors.