ContactUsLinkComponent
Solujet* Detergent
Solujet® Low-Foaming Phosphate-Free Liquid Detergents
# de catalogue CA80077-359
Fournisseur:  Alconox
Solujet* Detergent
Solujet® Low-Foaming Phosphate-Free Liquid Detergents
# de catalogue CA80077-359
Fournisseur:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    3.8 L (1 gal.)
  • Cat. no.:
    CA80077-359
  • pH:
    12
  • Supplier No.:
    2101
  • Packing Type:
    Bottle
Spécifications
A propos de cet article
Non ionic, low foaming phosphate free liquid cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. Passes inhibitory residue test.
  • Concentrated, biodegradable and readily disposable
  • Low-foaming high-wetting detergent for maximum performance
  • Ideal for CIP cleaning and floor standing lab washing machines
  • Dilute 1:200
For removeing soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals such as stainless steel. Solujet is inhibited for Aluminum. Clean-in place, laboratory warewashers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, and manual cleaning if proper skin and eye protection is used.
Note: Corrosion testing is advisable.
They can be used to clean pharmaceutical process equipment, water testing labware, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors.
Certifications: USDA authorized category A1 for use in food contact surface cleaning. FDA certified.
Customers Who Bought This Also Bought