PELTOR™ Optime™ Series Earmuffs, 3M™ Company
# de catalogue CA56615-921
Fournisseur: 3M
Spécifications
- NRR:25 dB (23 - 28 dB)
- Description:Over-the-head earmuffs Peltor™ Optime™ 98 with stainless steel headband
- Cat. no.:CA56615-921
- Supplier No.:H9A
Spécifications
A propos de cet article
Earmuffs combine excellent attenuation with comfortable durability. They provide a protective seal without excess pressure while the pivoting earcup connections help maintain the seal and proper alignment.
- Unique dual-cup design and soft, foam-filled cushions provide noise reduction and comfort
- Ultra-light with liquid/foam filled earmuff cushions
Double-shell technology boosts the noise blocking ability
Customers Who Bought This Also Bought