Spécifications
- Description:Corning® DMEM/Ham's F-12 50/50 Mix [+] L-glutamine
- Storage temperature:2...8 °C
- Media Format:Liquid
- Cat. no.:CA45000-344
- Shelf Life:12 months
- Supplier No.:10-090-CV
Spécifications
A propos de cet article
This 1:1 mixture of Dulbecco's Modified Eagle Medium (DMEM) and Ham's F-12 media is ideal for culture of a variety of cell types under low serum conditions
- Low endotoxin - <0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
DMEM is a modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME. Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids.