Nylaflo® Membrane Disc Filters
# de catalogue CA28140-041
Fournisseur: Cytiva
Spécifications
- Ø:90 mm
- Pore size:0.20 µm
- Cat. no.:CA28140-041
- Supplier No.:66603
Spécifications
A propos de cet article
Useful for a wide range of both aqueous and solvent-based applications.
Naturally wettable to aqueous solutions. HPLC certified for low levels of UV-absorbing extractables.
- Hydrophilic
- Excellent chemical compatibility
- Naturally wettable
Useful for a wide range of both aqueous and solvent-based applications.
Hydrophilic nylon filters offer excellent chemical compatibility with esters, bases, and alcohols
Naturally wettable to aqueous solutions. HPLC certified for low levels of UV-absorbing extractables.
Not recommended for acids >1N or halogenated solvents.
Customers Who Bought This Also Bought