ContactUsLinkComponent
Sterile Petri Dish, 50mm
Petri Dishes, Sterile, Cytiva (Formerly Pall Lab)
# de catalogue CA25388-606
Fournisseur:  Cytiva
Sterile Petri Dish, 50mm
Petri Dishes, Sterile, Cytiva (Formerly Pall Lab)
# de catalogue CA25388-606
Fournisseur:  Cytiva
Spécifications
  • Description:
    Plain
  • Cat. no.:
    CA25388-606
  • Packaging:
    Plastic Tray
  • Supplier No.:
    7242
Spécifications
A propos de cet article
PS dish, cellulose pad. Ideal for microbiological analysis when performing the membrane filter technique. Petri dishes with absorbent pads can be used with broth media, or users can pour agar into dishes without absorbent pads. Accepts 47 mm membrane filter.
  • Gamma irradiated so no Ethylene Oxide residuals to impede microbial growth
  • Easy to use, opens easily with one hand, yet closes to a tight seal
  • Uses less space on the laboratory bench or in an incubator with easy stacking base
Customers Who Bought This Also Bought