ContactUsLinkComponent
KIT - TEST 5B HARDNESS 0-30GPG
Total Hardness Test Kit, Model 5-B, Hach
# de catalogue CA1453-00
Fournisseur:  Hach
Numéro CAS:  
KIT - TEST 5B HARDNESS 0-30GPG
Total Hardness Test Kit, Model 5-B, Hach
# de catalogue CA1453-00
Fournisseur:  Hach
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    EDTA
  • Description:
    Total Hardness Test Kit, Model 5-B
  • Method:
    Drop count titration/ EDTA-bulk powder
  • Case style:
    L
  • Cat. no.:
    CA1453-00
  • Range:
    1 - 30 gpg CaCO₃
  • Supplier No.:
    1453-00
  • No. of Tests:
    100
Spécifications
A propos de cet article
Trust the original Hach test kit! Simple drop-count titration measures hardness as CaCO3 (1 gpg = 17.1 mg/L)
-Utilizes drop count titration - a simple, economical titration method
Utilizes drop count titration - a simple, economical titration method
Utilizes drop count titration with a range of 1-30 gpg.
Customers Who Bought This Also Bought