Spécifications
- Chemical method:EDTA
- Description:Total Hardness Test Kit, Model 5-B
- Method:Drop count titration/ EDTA-bulk powder
- Case style:L
- Cat. no.:CA1453-00
- Range:1 - 30 gpg CaCO₃
- Supplier No.:1453-00
- No. of Tests:100
Spécifications
A propos de cet article
Trust the original Hach test kit! Simple drop-count titration measures hardness as CaCO3 (1 gpg = 17.1 mg/L)
-Utilizes drop count titration - a simple, economical titration method
Utilizes drop count titration - a simple, economical titration method
Utilizes drop count titration with a range of 1-30 gpg.