Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:Silver
- Glove size:7
- Length:36.8 cm (14 3/8")
- Description:PE and EVOH
- Thickness:2.7 mil
- Cat. no.:CA11000-650
- Supplier No.:SSG/7
Spécifications
A propos de cet article
Gloves feature a five-layer fabric compacted to a thickness of 2.7mil.
Gloves feature a five-layer fabric compacted to a thickness of 2.7mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35°C. Each layer acts as a barrier against various epoxies and industrial chemicals.
Made of a unique, flexible film, Silver Shield®/4H® gloves can be worn by themselves or as permeation-resistant liners under other protective gloves. They are lightweight, ambidextrous, and allow for maximum worker protection, yet do not contain chemical accelerants that can cause allergic reactions. Smooth finish.
- Resist permeation of more than 280 chemicals
- Protect hands in chemical and petrochemical laboratories, spill cleanups, and other hazardous applications
- Ambidextrous, disposable, usable as secondary inner layer for maximum protection
Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35 °C
Made of a unique, flexible film, Silver Shield®/4H® gloves can be worn by themselves or as permeation-resistant liners under other protective gloves. They are lightweight, and allow for maximum worker protection, yet do not contain chemical accelerants that can cause allergic reactions. Smooth finish.
Each layer acts as a barrier against various epoxies and industrial chemicals.