ContactUsLinkComponent
AQUASTR CMBICLMT FRTLS BOTTLE CL500ML
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
# de catalogue CA1.09257.0500
Fournisseur:  MilliporeSigma
Numéro CAS:  
AQUASTR CMBICLMT FRTLS BOTTLE CL500ML
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
# de catalogue CA1.09257.0500
Fournisseur:  MilliporeSigma
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Packaging:
    Glass bottle
  • Description:
    Apura® CombiCoulomat Fritless Reagent
  • Size:
    500 ml
  • Cat. no.:
    CA1.09257.0500
  • Supplier No.:
    1.09257.0500
Spécifications
A propos de cet article
The Aquastar® range is designed to provide reagents for the determination of water by Karl Fischer titration of a wide range of substrates and by a variety of methodologies. These include reagents for one- or two-component systems; combined coulometric reagents; solvents for oils and fats; reagents and solvents for determination of water in ketones and aldehydes. Calibration standards are also available.
  • CombiTitrants and CombiSolvents are non toxic
  • Very accurate and reproducible results
  • Rapid and reliable
Merck have introduced into the range of Aquastar® Titrants, some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tags.
Customers Who Bought This Also Bought