ContactUsLinkComponent
SHARPS COLL 1 PIECE 1.4L
Chemotherapy Sharps Collectors, BD Medical
# de catalogue CA-BD300460A
Fournisseur:  BD
SHARPS COLL 1 PIECE 1.4L
Chemotherapy Sharps Collectors, BD Medical
# de catalogue CA-BD300460A
Fournisseur:  BD
Spécifications
  • Size:
    10 kg
  • Cat. no.:
    CA-BD300460A
  • Packaging:
    Pail
  • Supplier No.:
    300460
Spécifications
A propos de cet article
Units feature a wide entry port design for disposal of bulky chemotherapy waste
For accessories, see BD305451 series.
For accessories, see BD305451 series.
Collectors are designed to offer increased leak resistance. Units cannot be autoclaved. Plug cap provides a solid closure for leak resistant transit.
Customers Who Bought This Also Bought