ContactUsLinkComponent
BDH PH TEST STRIP 6.0-10.0
pH Test Strips, VWR Chemicals BDH®
Catalog # BDH83931.601
Supplier:  BDH
BDH PH TEST STRIP 6.0-10.0
pH Test Strips, VWR Chemicals BDH®
Catalog # BDH83931.601
Supplier:  BDH
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    BDH83931.601
  • pH Range:
    6.0 - 10.0
  • pH Graduation:
    0.3/0.5
  • Supplier No.:
    92122.2047
Specifications
About this item
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
  • Designed for safe testing of dangerous, poisonous, or aggressive liquids
  • Long handle protects user from contact with sample
  • Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Dimensions: (LxW) 85x6 mm.
Ordering information: Strips are packaged 100 to a box. Included color chart has 4 different color blocks for each pH value
Customers Who Bought This Also Bought