Spécifications
- pH Range:0.3 - 2.3
- pH Graduation:0.3/0.4
- Supplier No.:92180.2047
Spécifications
A propos de cet article
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
- Designed for safe testing of dangerous, poisonous, or aggressive liquids
- Long handle protects user from contact with sample
- Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
Dimensions: (LxW) 85x6 mm.